MilliporeSigma
  • Electrochemical Synthesis of Biaryls via Oxidative Intramolecular Coupling of Tetra(hetero)arylborates.
Electrochemical Synthesis of Biaryls via Oxidative Intramolecular Coupling of Tetra(hetero)arylborates.
Journal of the American Chemical Society (2020-02-11)
Arif Music, Andreas N Baumann, Philipp Spieß, Allan Plantefol, Thomas C Jagau, Dorian Didier
ABSTRACT
We report herein versatile, transition metal-free and additive-free (hetero)aryl-aryl coupling reactions promoted by the oxidative electrocoupling of unsymmetrical tetra(hetero)arylborates (TABs) prepared from ligand-exchange reactions on potassium trifluoroarylborates. Exploiting the power of electrochemical oxidations, this method complements the existing organoboron toolbox. We demonstrate the broad scope, scalability, and robustness of this unconventional catalyst-free transformation, leading to functionalized biaryls and ultimately furnishing drug-like small molecules, as well as late stage derivatization of natural compounds. In addition, the observed selectivity of the oxidative coupling reaction is related to the electronic structure of the TABs through quantum-chemical calculations and experimental investigations.