MilliporeSigma
  • --Essential-oil composition of the fruits of six Heracleum L. species from Iran: chemotaxonomic significance.
--Essential-oil composition of the fruits of six Heracleum L. species from Iran: chemotaxonomic significance.
Chemistry & biodiversity (2014-12-11)
Tayebeh Radjabian, Azam Salimi, Nosrat Rahmani
ABSTRACT
The fruit essential oils of Heracleum persicum, H. rechingeri, H. gorganicum, H. rawianum, H. pastinacifolium, and H. anisactis from Iran were obtained by hydrodistillation and characterized by GC-FID and GC/MS analyses. The oils of the six species were compared to determine the similarities and differences among their compositions. Overall, 36 compounds were identified in the fruit oils, accounting for 92.40-96.74% of the total oil compositions. Aliphatic esters constituted the main fraction of the oils (86.61-94.31%), with octyl acetate and hexyl butyrate as the major components. The oil compositions of species belonging to section Pubescentia (H. persicum, H. gorganicum, and H. rechingeri) were discriminated by equally high contents of both octyl acetate (13.84-20.48%) and hexyl butyrate (17.73-38.36%). On the other hand, the oils of H. rawianum, H. pastinacifolium and H. anisactis, belonging to section Wendia, showed lower hexyl butyrate contents (3.62-6.6%) and higher octyl acetate contents (48.71-75.36%) than the former. Moreover, isoelemicin was identified at low amounts (0.10-2.51%) only in the oils of the latter species. The differences in the oil composition among the six species were investigated by hierarchical cluster and principal component analyses, which indicated that the oil composition confirmed well the taxonomical classification based on the morphological and botanical data, and, thus, may provide a reliable marker to discriminate Heracleum species at the intersectional level.