MilliporeSigma
  • A new entry to Amaryllidaceae alkaloids from carbohydrates: total synthesis of (+)-vittatine.
A new entry to Amaryllidaceae alkaloids from carbohydrates: total synthesis of (+)-vittatine.
Chemical communications (Cambridge, England) (2004-04-30)
Masahiro Bohno, Hidetomo Imase, Noritaka Chida
ABSTRACT
The stereoselective and chiral synthesis of the Amaryllidaceae alkaloid, (+)-vittatine 1, is described; the quaternary carbon in 1 was generated by Claisen rearrangement of a cyclohexenol derived from D-glucose by way of a Ferrier's carbocyclisation reaction and a hexahydroindole skeleton was effectively constructed by intramolecular aminomercuration-demercuration, followed by Chugaev reaction.