MilliporeSigma
  • Zafirlukast promotes insulin secretion by increasing calcium influx through L-type calcium channels.
Zafirlukast promotes insulin secretion by increasing calcium influx through L-type calcium channels.
Journal of cellular physiology (2018-05-26)
Hyeon-Jeong Hwang, Kyoung-Su Park, Jang Hyun Choi, Lucio Cocco, Hyun-Jun Jang, Pann-Ghill Suh
ABSTRACT
The zafirlukast has been reported to be anti-inflammatory and widely used to alleviate the symptoms of asthma. However, its influence on insulin secretion in pancreatic β-cells has not been investigated. Herein, we examined the effects of zafirlukast on insulin secretion and the potential underlying mechanisms. Among the cysteinyl leukotriene receptor 1 antagonists, zafirlukast, pranlukast, and montelukast, only zafirlukast enhanced insulin secretion in a concentration-dependent manner in both low and high glucose conditions and elevated the level of [Ca2+ ]i , further activating Ca2+ /calmodulin-dependent protein kinase II (CaMKII), protein kinase B (AKT), and extracellular signal-regulated kinase (ERK) signaling. These effects were nearly abolished by the L-type Ca2+ channel antagonist nifedipine, while treatment with thapsigargin, a sarco/endoplasmic reticulum Ca2+ ATPase inhibitor, did not have the same effect, suggesting that zafirlukast primarily induces the entry of extracellular Ca2+ rather than intracellular Ca2+ from the endoplasmic reticulum. Zafirlukast treatment resulting in a significant drop in glucose levels and increased insulin secretion in C57BL/6J mice. These findings will contribute to an improved understanding of the side effects of zafirlukast and potential candidate for a therapeutic intervention in diabetes.
Sigma-Aldrich
Zafirlukast, ≥98% (HPLC)