MilliporeSigma
  • The GOLD domain-containing protein TMED7 inhibits TLR4 signalling from the endosome upon LPS stimulation.
The GOLD domain-containing protein TMED7 inhibits TLR4 signalling from the endosome upon LPS stimulation.
Nature communications (2012-03-20)
Sarah L Doyle, Harald Husebye, Dympna J Connolly, Terje Espevik, Luke A J O'Neill, Anne F McGettrick
ABSTRACT
Toll-like receptor 4 is an innate immune receptor responsible for the recognition of the Gram-negative cell wall component lipopolysaccharide. Here we show that transmembrane emp24 domain-containing protein 7 (TMED7) inhibits MyD88-independent toll-like receptor 4 signalling. TMED7 overexpression inhibits the ability of TRAM, an adaptor utilized by toll-like receptor 4, or lipopolysaccharide to activate the interferon regulatory factor 3-signalling pathway, whereas TMED7 knockdown enhances production of the cytokine, RANTES, following lipopolysaccharide stimulation. Upon lipopolysaccharide stimulation, TMED7 co-localizes with TRAM and toll-like receptor 4 in late endosomes where it encounters the negative regulator of TRAM, TAG. The TMED7 sequence is found in TAG because of a read-through from the tmed7 gene into the ticam2 gene. TMED7 is essential for TAG-mediated disruption of the TRAM/TRIF complex and the degradation of toll-like receptor 4. A TMED homologue, logjam, has a negative role in the Toll and IMD pathways in Drosophila melanogaster; therefore, TMEDs may have a conserved role in the regulation of innate immunity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.