MilliporeSigma
  • Indoor use of attractive toxic sugar bait in combination with long-lasting insecticidal net against pyrethroid-resistant Anopheles gambiae: an experimental hut trial in Mbé, central Côte d'Ivoire.
Indoor use of attractive toxic sugar bait in combination with long-lasting insecticidal net against pyrethroid-resistant Anopheles gambiae: an experimental hut trial in Mbé, central Côte d'Ivoire.
Malaria journal (2020-01-09)
Joanna E C Furnival-Adams, Soromane Camara, Mark Rowland, Alphonsine A Koffi, Ludovic P Ahoua Alou, Welbeck A Oumbouke, Raphael N'Guessan
ABSTRACT
Indoor attractive toxic sugar bait (ATSB) has potential as a supplementary vector-control and resistance-management tool, offering an alternative mode of insecticide delivery to current core vector-control interventions, with potential to deliver novel insecticides. Given the high long-lasting insecticidal bed net (LLIN) coverage across Africa, it is crucial that the efficacy of indoor ATSB in combination with LLINs is established before it is considered for wider use in public health. An experimental hut trial to evaluate the efficacy of indoor ATSB traps treated with 4% boric acid (BA ATSB) or 1% chlorfenapyr (CFP ATSB) in combination with untreated nets or LLINs (holed or intact), took place at the M'bé field station in central Côte d'Ivoire against pyrethroid resistant Anopheles gambiae sensu lato. The addition of ATSB to LLINs increased the mortality rates of wild pyrethroid-resistant An. gambiae from 19% with LLIN alone to 28% with added BA ATSB and to 39% with added CFP ATSB (p < 0.001). Anopheles gambiae mortality with combined ATSB and untreated net was similar to that of combined ATSB and LLIN regardless of which insecticide was used in the ATSB. The presence of holes in the LLIN did not significantly affect ATSB-induced An. gambiae mortality. Comparative tests against pyrethroid resistant and susceptible strains using oral application of ATSB treated with pyrethroid demonstrated 66% higher survival rate among pyrethroid-resistant mosquitoes. Indoor ATSB traps in combination with LLINs enhanced the control of pyrethroid-resistant An. gambiae. However, many host-seeking An. gambiae entering experimental huts with indoor ATSB exited into the verandah trap without sugar feeding when restricted from a host by a LLIN. Although ATSB has potential for making effective use of classes of insecticide otherwise unsuited to vector control, it does not exempt potential selection of resistance via this route.
Product Description
Supelco
Chlorfenapyr, PESTANAL®, analytical standard