MilliporeSigma
  • Lifetime-based sensing of the hyaluronidase using fluorescein labeled hyaluronic acid.
Lifetime-based sensing of the hyaluronidase using fluorescein labeled hyaluronic acid.
Journal of photochemistry and photobiology. B, Biology (2011-11-16)
Rafal Fudala, Mark E Mummert, Zygmunt Gryczynski, Ryan Rich, Julian Borejdo, Ignacy Gryczynski
ABSTRACT
In this report we propose a lifetime-based sensing (LBS) for the detection of hyaluronidase (HA-ase). First, we heavily label hyaluronan macromolecules (HAs) with fluorescein amine. The fluorescein labeled HA (HA-Fl) has a weak fluorescence and short fluorescence lifetime due to an efficient self-quenching. Upon the addition of HA-ase, the brightness and lifetime of the sample increase. The cleavage of an HA macromolecule reduces the energy migration between fluorescein molecules and the degree of the self-quenching. A first order of the cleavage reaction depends on the amount of the HA-ase enzyme. We describe an HA-ase sensing strategy based on the lifetime changes of the fluorescein labeled HA in the presence of HA-ase. We demonstrate that the calibration of the sensing response is the same for the average lifetime as for a single exponential decay approximation, which significantly simplifies the analysis of the sensing measurements.
Sigma-Aldrich
Fluorescein hyaluronic acid, >95%, powder