MilliporeSigma
  • A new strategy based on gas chromatography-high resolution mass spectrometry (GC-HRMS-Q-Orbitrap) for the determination of alkenylbenzenes in pepper and its varieties.
A new strategy based on gas chromatography-high resolution mass spectrometry (GC-HRMS-Q-Orbitrap) for the determination of alkenylbenzenes in pepper and its varieties.
Food chemistry (2020-04-14)
Araceli Rivera-Pérez, Rosalía López-Ruiz, Roberto Romero-González, Antonia Garrido Frenich
ABSTRACT
Alkenylbenzenes are natural toxins with genotoxic and carcinogenic effects in rodents, which are highly present in condiments frequently consumed. The aim of this study was the development of the first multi-analyte method for the determination of eight alkenylbenzenes (eugenol, methyl eugenol, acetyl eugenol, trans-isoeugenol, safrole, estragole, myristicin and trans-anethole) in different pepper varieties by gas chromatography coupled to high-resolution mass spectrometry (GC-HRMS-Q-Orbitrap) in combination with a simple ultrasound-assisted extraction method (UAE). The method was successfully validated, and it was applied for studying the presence of these analytes in peppers as well as to elucidate the effects of the berries' maturity and the geographical origin on alkenylbenzene contents. The analysis of the pepper samples showed that eugenol (10.5-120 mg/kg), trans-anethole (10.7-42.7 mg/kg) and estragole (2.2-45.7 mg/kg) tended to be the most detected alkenylbenzenes at high levels, whereas trans-isoeugenol (0.69-3.6 mg/kg) and safrole (0.20-3.0 mg/kg) were minor components. Estragole (PubChem CID: 8815); trans-anethole (PubChem CID: 637563); Myristicin (PubChem CID: 4276); Safrole (PubChem CID: 5144); Eugenol (PubChem CID: 3314); Methyl eugenol (PubChem CID: 7127); Acetyl eugenol (PubChem CID: 7136); trans-Isoeugenol (PubChem CID: 853433); Caffeine (PubChem CID: 2519); Dicyclohexylmethanol (PubChem CID: 78197).