MilliporeSigma
  • Neuromedin U type 1 receptor stimulation of A-type K+ current requires the βγ subunits of Go protein, protein kinase A, and extracellular signal-regulated kinase 1/2 (ERK1/2) in sensory neurons.
Neuromedin U type 1 receptor stimulation of A-type K+ current requires the βγ subunits of Go protein, protein kinase A, and extracellular signal-regulated kinase 1/2 (ERK1/2) in sensory neurons.
The Journal of biological chemistry (2012-04-12)
Yiming Zhang, Dongsheng Jiang, Yuan Zhang, Xinghong Jiang, Fen Wang, Jin Tao
ABSTRACT
Although neuromedin U (NMU) has been implicated in analgesia, the detailed mechanisms still remain unclear. In this study, we identify a novel functional role of NMU type 1 receptor (NMUR1) in regulating the transient outward K(+) currents (I(A)) in small dorsal root ganglion (DRG) neurons. We found that NMU reversibly increased I(A) in a dose-dependent manner, instead the sustained delayed rectifier K(+) current (I(DR)) was not affected. This NMU-induced I(A) increase was pertussis toxin-sensitive and was totally reversed by NMUR1 knockdown. Intracellular application of GDPβS (guanosine 5'-O-(2-thiodiphosphate)), QEHA peptide, or a selective antibody raised against the Gα(o) or Gβ blocked the stimulatory effects of NMU. Pretreatment of the cells with the protein kinase A (PKA) inhibitor or ERK inhibitor abolished the NMU-induced I(A) response, whereas inhibition of phosphatidylinositol 3-kinase or PKC had no such effects. Exposure of DRG neurons to NMU markedly induced the phosphorylation of ERK (p-ERK), whereas p-JNK or p-p38 was not affected. Moreover, the NMU-induced p-ERK increase was attenuated by PKA inhibition and activation of PKA by foskolin would mimic the NMU-induced I(A) increase. Functionally, we observed a significant decrease of the firing rate of neuronal action potential induced by NMU and pretreatment of DRG neurons with 4-AP could abolish this effect. In summary, these results suggested that NMU increases I(A) via activation of NMUR1 that couples sequentially to the downstream activities of Gβγ of the G(o) protein, PKA, and ERK, which could contribute to its physiological functions including neuronal hypoexcitability in DRG neurons.
Sigma-Aldrich
3-(2-Aminoethyl)-5-((4-ethoxyphenyl)methylene)-2,4-thiazolidinedione hydrochloride, powder, ≥98% (HPLC)
Pricing and availability is not currently available.