MilliporeSigma
  • FGF6 enhances muscle regeneration after nerve injury by relying on ERK1/2 mechanism.
FGF6 enhances muscle regeneration after nerve injury by relying on ERK1/2 mechanism.
Life sciences (2020-02-28)
Qiuchen Cai, Genbin Wu, Min Zhu, Heng''an Ge, Chao Xue, Qing''gang Zhang, Biao Cheng, Sudan Xu, Peng Wu
ABSTRACT
Severe peripheral nerve injury leads to skeletal muscle atrophy and impaired limb function that is not sufficiently improved by existing treatments. Fibroblast growth factor 6 (FGF6) is involved in tissue regeneration and is dysregulated in denervated rat muscles. However, the way that FGF6 affects skeletal muscle repair after peripheral nerve injury has not been fully elucidated. In this study, we investigated the role of FGF6 in the regeneration of denervated muscles using myoblast cells and an in vivo model of peripheral nerve injury. FGF6 promoted the viability and migration of C2C12 and primary myoblasts in a dose-dependent manner through FGFR1-mediated upregulation of cyclin D1. Low concentrations of FGF6 promoted myoblast differentiation through FGFR4-mediated activation of ERK1/2, which upregulated expression of MyHC, MyoD, and myogenin. FGFR-1, FGFR4, MyoD, and myogenin were not upregulated when FGF6 expression was inhibited in myoblasts by shRNA-mediated knockdown. Injection of FGF6 into denervated rat muscles enhanced the MyHC-IIb muscle fiber phenotype and prevented muscular atrophy. These findings indicate that FGF6 reduces skeletal muscle atrophy by relying on the ERK1/2 mechanism and enhances the conversion of slow muscle to fast muscle fibers, thereby promoting functional recovery of regenerated skeletal muscle after innervation.
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR4
Pricing and availability is not currently available.