MilliporeSigma
Retinol-binding protein 4, fetal overgrowth and fetal growth factors.
Pediatric research (2019-12-01)
Meng-Nan Yang, Guang-Hui Zhang, Kun Du, Wen-Juan Wang, Yu Dong, Hua He, Huei-Chen Chiu, Yu-Na Guo, Fengxiu Ouyang, Jun Zhang, Xiao-Lin Hua, Zhong-Cheng Luo
ABSTRACT
Retinol-binding protein 4 (RBP-4) is an adipokine involved in regulating insulin sensitivity which would affect fetal growth. It is unclear whether RBP-4 is associated with fetal overgrowth, and unexplored which fetal growth factor(s) may mediate the association. In the Shanghai Birth Cohort, we studied 125 pairs of larger-for-gestational-age (LGA, birth weight >90th percentile, an indicator of fetal overgrowth) and optimal-for-gestational-age (OGA, 25-75th percentiles) control infants matched by sex and gestational age. We measured cord blood concentrations of RBP-4, insulin, proinsulin, insulin-like growth factor-I (IGF-I), and IGF-II. Cord blood RBP-4 concentrations were elevated in LGA vs. OGA infants (21.9 ± 6.2 vs. 20.2 ± 5.1 µg/ml, P = 0.011), and positively correlated with birth weight z score (r = 0.19, P = 0.003), cord blood proinsulin (r = 0.21, P < 0.001), IGF-I (r = 0.24, P < 0.001), and IGF-II (r = 0.15, P = 0.016). Adjusting for maternal and neonatal characteristics, each SD increment in cord blood RBP-4 was associated with a 0.28 (0.12-0.45) increase in birth weight z score (P < 0.001). Mediation analyses showed that IGF-I could account for 31.7% of the variation in birth weight z score in association with RBP-4 (P = 0.01), while IGF-II was not an effect mediator. RBP-4 was positively associated with fetal overgrowth. IGF-I (but not IGF-II) may mediate this association.
Sigma-Aldrich
Mouse IGF-II ELISA Kit, for cell culture supernatant
Pricing and availability is not currently available.
Sigma-Aldrich
Bovine INS / Insulin ELISA Kit
Pricing and availability is not currently available.