MilliporeSigma
  • Potential for release of pulmonary toxic ketene from vaping pyrolysis of vitamin E acetate.
Potential for release of pulmonary toxic ketene from vaping pyrolysis of vitamin E acetate.
Proceedings of the National Academy of Sciences of the United States of America (2020-03-12)
Dan Wu, Donal F O'Shea
ABSTRACT
A combined analytical, theoretical, and experimental study has shown that the vaping of vitamin E acetate has the potential to produce exceptionally toxic ketene gas, which may be a contributing factor to the upsurge in pulmonary injuries associated with using e-cigarette/vaping products. Additionally, the pyrolysis of vitamin E acetate also produces carcinogen alkenes and benzene for which the negative long-term medical effects are well recognized. As temperatures reached in vaping devices can be equivalent to a laboratory pyrolysis apparatus, the potential for unexpected chemistries to take place on individual components within a vape mixture is high. Educational programs to inform of the danger are now required, as public perception has grown that vaping is not harmful.
Sigma-Aldrich
Aldrich® low form dewar flask, capacity 150 mL, hemispherical, depth 4 cm
Sigma-Aldrich
(+)-α-Tocopherol acetate, BioReagent, suitable for insect cell culture, ~1360 IU/g