MilliporeSigma
  • Early Aspirin Discontinuation Following Acute Coronary Syndrome or Percutaneous Coronary Intervention: A Systematic Review and Meta-Analysis of Randomized Controlled Trials.
Early Aspirin Discontinuation Following Acute Coronary Syndrome or Percutaneous Coronary Intervention: A Systematic Review and Meta-Analysis of Randomized Controlled Trials.
Journal of clinical medicine (2020-03-07)
Paul Guedeney, Jules Mesnier, Sabato Sorrentino, Farouk Abcha, Michel Zeitouni, Benoit Lattuca, Johanne Silvain, Salvatore De Rosa, Ciro Indolfi, Jean-Philippe Collet, Mathieu Kerneis, Gilles Montalescot
ABSTRACT
The respective ischemic and bleeding risks of early aspirin discontinuation following an acute coronary syndrome (ACS) or percutaneous coronary intervention (PCI) remain uncertain. We performed a prospero-registered review of randomized controlled trials (RCTs) comparing a P2Y12 inhibitor-based single antiplatelet strategy following early aspirin discontinuation to a strategy of sustained dual antiplatelet therapy (DAPT) in ACS or PCI patients requiring, or not, anticoagulation for another indication (CRD42019139576). We estimated risk ratios (RR) and 95% confidence intervals (CI) using random effect models. We included nine RCTs comprising 40,621 patients. Compared to prolonged DAPT, major bleeding (2.2% vs. 2.8%; RR 0.68; 95% CI: 0.54 to 0.87; p = 0.002; I²: 63%), non-major bleeding (5.0 % vs. 6.1 %; RR: 0.66; 95% CI: 0.47 to 0.94; p = 0.02; I² : 87%) and all bleeding (7.4% vs. 9.9%; RR: 0.65; 95% CI: 0.53 to 0.79; p < 0.0001; I²: 88%) were significantly reduced with early aspirin discontinuation without significant difference for all-cause death (p = 0.60), major adverse cardiac and cerebrovascular events (MACE) (p = 0.60), myocardial infarction (MI) (p = 0.77), definite stent thrombosis (ST) (p = 0.63), and any stroke (p = 0.59). In patients on DAPT after an ACS or a PCI, early aspirin discontinuation prevents bleeding events with no significant adverse effect on the ischemic risk or mortality.
Sigma-Aldrich
NCX 4040, ≥98% (HPLC)