MilliporeSigma
  • Low birth weight influences the postnatal abundance and characteristics of satellite cell subpopulations in pigs.
Low birth weight influences the postnatal abundance and characteristics of satellite cell subpopulations in pigs.
Scientific reports (2020-04-11)
K Stange, C Miersch, G Sponder, M Röntgen
ABSTRACT
Low birth weight (LBW) can cause lifelong impairments in muscle development and growth. Satellite cells (SC) and their progeny are crucial contributors to myogenic processes. This study provides new data on LBW in piglets combining insights on energy metabolism, muscle capillarization and differences in SC presence and function. To this aim, muscle tissues as well as isolated myogenic cells of 4-day-old German Landrace piglets were analyzed. For the first time two heterogeneous SC subpopulations, which contribute differently to muscle development, were isolated from LBW pigs by Percoll density gradient centrifugation. The muscles of LBW piglets showed a reduced DNA, RNA, and protein content as well as lower activity of the muscle specific enzymes CK, ICDH, and LDH compared to their normal birth weight siblings. We assume that deficits in energy metabolism and capillarization are associated with reduced bioavailability of SC, possibly leading to early exhaustion of the SC reserve cell pool and the cells' premature differentiation.
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%