MilliporeSigma
  • The Effects of Salts and Osmoprotectants on Enzyme Activities of Fructose-1,6-biphosphate Aldolases in a Halotolerant Cyanobacterium, Halothece sp. PCC 7418.
The Effects of Salts and Osmoprotectants on Enzyme Activities of Fructose-1,6-biphosphate Aldolases in a Halotolerant Cyanobacterium, Halothece sp. PCC 7418.
Life (Basel, Switzerland) (2020-03-19)
Siripat Ngoennet, Masaki Honda, Tanutcha Patipong, Takashi Hibino, Rungaroon Waditee-Sirisattha, Hakuto Kageyama
ABSTRACT
The halotolerant cyanobacterium, Halothece sp. PCC 7418, possesses two classes of fructose-1,6-bisphosphate aldolase (FBA): H2846 and H2847. Though class I (CI)-FBA H2846 is thought to be associated with salt tolerance, the regulatory mechanisms, molecular characteristics, and expression profiles between H2846 and class II (CII)-FBA H2847 have scarcely been investigated. Here, we show that the accumulation of the H2846 protein is highly responsive to both up- and down-shock with NaCl, whereas H2847 is constitutively expressed. The activity of CI- and CII-FBA in cyanobacterial extracts is correlated with the accumulation patterns of H2846 and H2847, respectively. In addition, it was found that these activities were inhibited by NaCl and KCl, with CII-FBA activity strikingly inhibited. It was also found that the CI-FBA activity of recombinant H2846 was hindered by salts and that this hindrance could be moderated by the addition of glycine betaine (GB), whereas no moderation occurred with other potential osmoprotectant molecules (proline, sucrose, and glycerol). In addition, a phylogenetic analysis showed that CI-FBAs with higher similarities to H2846 tended to be distributed among potential GB-synthesizing cyanobacteria. Taken together, our results provide insights into the independent evolution of the CI- and CII-FBA gene families, which show distinct expression profiles and functions following salt stress.
Sigma-Aldrich
D-Glyceraldehyde 3-phosphate solution, 8-13 mg/mL in H2O