MilliporeSigma
  • Determination of chondroitin sulfate content in raw materials and dietary supplements by high-performance liquid chromatography with ultraviolet detection after enzymatic hydrolysis: single-laboratory validation.
Determination of chondroitin sulfate content in raw materials and dietary supplements by high-performance liquid chromatography with ultraviolet detection after enzymatic hydrolysis: single-laboratory validation.
Journal of AOAC International (2007-06-22)
David Ji, Mark Roman, Joseph Zhou, Jana Hildreth
ABSTRACT
A method to quantify chondroitin sulfate in raw materials and dietary supplements at a range of about 5 to 100% (w/w) chondroitin sulfate has been developed and validated. The chondroitin sulfate is first selectively hydrolyzed by chondroitinase ACII enzyme to form un-, mono-, di-, and trisulfated unsaturated disaccharides; the resulting disaccharides are then quantified by ion-pairing liquid chromatography with ultraviolet detection. The amounts of the individual disaccharides are summed to yield the total amount of chondroitin sulfate in the material. Single-laboratory validation has been performed to determine the repeatability, accuracy, selectivity, limit of detection, limit of quantification, ruggedness, and linearity of the method. Repeatability precision for total chondroitin sulfate content was between 1.60 and 4.72% relative standard deviation, with HorRat values between 0.79 and 2.25. Chondroitin sulfate recovery from raw material negative control was between 101 and 102%, and recovery from finished product negative control was between 105 and 106%.
Product Description
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Chondroitin sulfate sodium salt from bovine cartilage, 98-102% (CPC, titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Chondroitinase AC from Flavobacterium heparinum, lyophilized powder, 0.5-1.5 units/mg protein (using chondroitin sulfate A as substrate)
Pricing and availability is not currently available.