MilliporeSigma
  • Synthesis and characterisation of thiobarbituric acid enamine derivatives, and evaluation of their α-glucosidase inhibitory and anti-glycation activity.
Synthesis and characterisation of thiobarbituric acid enamine derivatives, and evaluation of their α-glucosidase inhibitory and anti-glycation activity.
Journal of enzyme inhibition and medicinal chemistry (2020-03-12)
M Ali, Assem Barakat, Ayman El-Faham, Hessa H Al-Rasheed, Kholoud Dahlous, Abdullah Mohammed Al-Majid, Anamika Sharma, Sammer Yousuf, Mehar Sanam, Zaheer Ul-Haq, M Iqbal Choudhary, Beatriz G de la Torre, Fernando Albericio
ABSTRACT
A new series of thiobarbituric (thiopyrimidine trione) enamine derivatives and its analogues barbituric acid derivatives was synthesised, characterised, and screen for in vitro evaluation of α-glucosidase enzyme inhibition and anti-glycation activity. This series of compounds were found to inhibit α-glucosidase activity in a reversible mixed-type manner with IC50 between 264.07 ± 1.87 and 448.63 ± 2.46 µM. Molecular docking studies indicated that compounds of 3g, 3i, 3j, and 5 are located close to the active site of α-glucosidase, which may cover the active pocket, thereby inhibiting the binding of the substrate to the enzyme. Thiopyrimidine trione derivatives also inhibited the generation of advanced glycation end-products (AGEs), which cause long-term complications in diabetes. While, compounds 3a-k, 5, and 6 showed significant to moderate anti-glycation activity (IC50 = 31.5 ± 0.81 to 554.76 ± 9.1 µM).
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Pricing and availability is not currently available.