MilliporeSigma
  • Fibronectin acts as a molecular switch to determine SPARC function in pancreatic cancer.
Fibronectin acts as a molecular switch to determine SPARC function in pancreatic cancer.
Cancer letters (2020-03-03)
Amanda Munasinghe, Khalisha Malik, Fatemia Mohamedi, Stan Moaraf, Hemant Kocher, Lucy Jones, Natasha J Hill
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) is one of the deadliest of all solid tumours and more effective therapy is urgently needed. The stroma is thought to play a critical role in tumour development and metastasis, and high stromal expression of the matricellular protein SPARC has been robustly associated with poor patient prognosis. However, the precise role of SPARC has been highly controversial, with multiple studies demonstrating tumour-suppressor properties of this protein in vitro. This conflicting data has been a barrier to the development of new therapeutic approaches targeting SPARC, despite current interest in stromal-therapy. We show conclusively that SPARC acts directly on cancer cells to promote pancreatic cancer cell proliferation. This contradicts previous in vitro studies, but is consistent with the observed clinical association between SPARC expression and poor patient prognosis. However, depletion of fibronectin switches the activity of SPARC from promoting cancer cell proliferation to growth inhibition and induction of apoptosis. Thus, targeting the interaction between SPARC and fibronectin could be used to turn the highly expressed tumour protein SPARC against the tumour to induce tumour cytotoxicity, and is a novel target for PDAC therapy.
Sigma-Aldrich
Human Fibronectin ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
Bovine Fibronectin ELISA Kit, for cell culture supernatants, plasma, and serum samples