MilliporeSigma
  • Addressing the challenge of changing the specificity of RNase T1 with rational and evolutionary approaches.
Addressing the challenge of changing the specificity of RNase T1 with rational and evolutionary approaches.
Chembiochem : a European journal of chemical biology (2004-02-05)
Marc Struhalla, Rico Czaja, Ulrich Hahn
ABSTRACT
Although ribonuclease T1 (RNase T1) is one of the best-characterized proteins with respect to structure and enzymatic action, numerous attempts at altering the specificity of the enzyme to cleave single-stranded RNA at the 3'-side of adenylic instead of guanylic residues by rational approaches have failed so far. Recently we generated and characterized the RNase T1 variant RV with a 7200-fold increase in adenylyl-3',5'-cytidine (ApC)/guanylyl-3',5'-cytidine (GpC) preference, with the guanine-binding loop changed from 41-KYNNYE-46 (wt) to 41-EFRNWN-46. Now we have introduced the asparagine residue at position 46 of the wild-type enzyme as a single-point mutation in variant E46N and in combination with the Y45W exchange also occurring in RV. Both variants show an improved ApC/GpC preference with a 1450-fold increase for E46N and a 2100-fold increase for Y45W/E46N in comparison to wild-type activity. We also addressed the challenge of altering enzyme specificity with an evolutionary approach. We have randomly introduced point mutations into the RNase T1 wild-type gene and into the gene of the variant RV with different mutation rates. Altogether we have screened about 100,000 individual clones for activity on RNase indicator plates; 533 of these clones were active. A significant change in substrate specificity towards an ApC preference could not be observed for any of these active variants; this demonstrated the magnitude of the challenge to alter the specificity of this evolutionary perfected enzyme.