MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
An enzymatic method for the detection of human serum albumin.
Experimental & molecular medicine (2001-07-20)
M U Javed, S N Waqar
ABSTRACT
Albumin is the most abundant protein in human serum. A dye-binding method is commonly used in clinical laboratories for its estimation using different types of dyes. However, all these dye methods were interfered by a variety of compounds. Here we present a method for the detection of albumin in human serum and other biological fluids. The principle is based on the fact that lactate dehydrogenase isoenzyme-5 (LDH-5) binds specifically to Dextran-Blue (DB). Albumin inhibits the binding of LDH-5 with DB. Absence of LDH activity in DB fraction after gel filtration indicates the presence of albumin in sample and vice versa.