MilliporeSigma
  • An LC/MS/MS method for quantitation of chemopreventive sphingadienes in food products and biological samples.
An LC/MS/MS method for quantitation of chemopreventive sphingadienes in food products and biological samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-08-05)
J H Suh, A M Makarova, J M Gomez, L A Paul, J D Saba
ABSTRACT
Colorectal cancer (CRC) is a leading cause of cancer mortality. Diet has a significant influence on colon cancer risk. Identifying chemopreventive agents, dietary constituents, practices and/or diet supplements that promote gut health and reduce the incidence of intestinal neoplasias and CRC could significantly impact public health. Sphingadienes (SDs) are dietary sphingolipids found in plant-based food products. SDs are cytotoxic to colon cancer cells and exhibit chemopreventive properties. The aim of the present study was to develop a sensitive and robust ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) method for quantifying SDs in food products and biological samples. The assay was linear over a concentration range of 80nM to 50μM and was sensitive to a detection limit of 3.3nM. Post-extraction stability was 100% at 24h. SD content in soy oils was approximately 10nM. SDs were detected transiently in the plasma of adult mice 10min after gavage delivery of a 25mg/kg bolus and declined to baseline by 1h. SD uptake in the gut was maximal in the duodenum and peaked 1h after gavage delivery. Disappearance of SDs in the lower gastrointestinal tract suggests either rapid metabolism to yet unidentified products or potentially luminal export.