MilliporeSigma
  • Oxysterol regulation of estrogen receptor alpha-mediated gene expression in a transcriptional activation assay system using HeLa cells.
Oxysterol regulation of estrogen receptor alpha-mediated gene expression in a transcriptional activation assay system using HeLa cells.
Bioscience, biotechnology, and biochemistry (2004-08-24)
Hiroyoshi Sato, Satoko Nishida, Hiroko Tomoyori, Masao Sato, Ikuo Ikeda, Katsumi Imaizumi
ABSTRACT
In order to test the estrogenic activity of sterol oxidation products from cholesterol and phytosterols, an estrogen-dependent gene expression assay was performed in estrogen receptor alpha-stably transformed HeLa cells. The ranking of the estrogenic potency of these compounds was different: 17beta-estradiol > genistein > beta-epoxycholesterol = daidzein = cholestanetriol = 22(R)-hydroxycholesterol = 20(S)-hydroxycholesterol = sitostanetriol > campestanetriol = beta-epoxysitosterol = 7beta-hydroxycholesterol. These compounds were not estrogenic in estrogen receptor-negative HeLa cells.