MilliporeSigma
  • Plasma membrane targeting by short chain sphingolipids inserted in liposomes improves anti-tumor activity of mitoxantrone in an orthotopic breast carcinoma xenograft model.
Plasma membrane targeting by short chain sphingolipids inserted in liposomes improves anti-tumor activity of mitoxantrone in an orthotopic breast carcinoma xenograft model.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-05-20)
Lília R Cordeiro Pedrosa, Olaf van Tellingen, Thomas Soullié, Ann L Seynhaeve, Alexander M M Eggermont, Timo L M Ten Hagen, Marcel Verheij, Gerben A Koning
ABSTRACT
Mitoxantrone (MTO) is clinically used for treatment of various types of cancers providing an alternative for similarly active, but more toxic chemotherapeutic drugs such as anthracyclines. To further decrease its toxicity MTO was encapsulated into liposomes. Although liposomal drugs can accumulate in target tumor tissue, they still face the plasma membrane barrier for effective intracellular delivery. Aiming to improve MTO tumor cell availability, we used short chain lipids to target and modulate the tumor cell membrane, promoting MTO plasma membrane traversal. MTO was encapsulated in liposomes containing the short chain sphingolipid (SCS), C8-Glucosylceramide (C8-GluCer) or C8-Galactosylceramide (C8-GalCer) in their bilayer. These new SCS-liposomes containing MTO (SCS-MTOL) were tested in vivo for tolerability, pharmacokinetics, biodistribution, tumor drug delivery by intravital microscopy and efficacy, and compared to standard MTO liposomes (MTOL) and free MTO. Liposomal encapsulation decreased MTO toxicity and allowed administration of higher drug doses. SCS-MTOL displayed increased clearance and lower skin accumulation compared to standard MTOL. Intratumoral liposomal drug delivery was heterogeneous and rather limited in hypoxic tumor areas, yet SCS-MTOL improved intracellular drug uptake in comparison with MTOL. The increased MTO availability correlated well with the improved antitumor activity of SCS-MTOL in a MDAMB-231 breast carcinoma model. Multiple dosing of liposomal MTO strongly delayed tumor growth compared to free MTO and prolonged mouse survival, whereas among the liposomal MTO treatments, C8-GluCer-MTOL was most effective. Targeting plasma membranes with SCS improved MTO tumor availability and thereby therapeutic activity and represents a promising approach to improve MTO-based chemotherapy.