MilliporeSigma
  • Inhibitors of acyl-CoA:cholesterol acyltransferase (ACAT). 7. Development of a series of substituted N-phenyl-N'-[(1-phenylcyclopentyl)methyl]ureas with enhanced hypocholesterolemic activity.
Inhibitors of acyl-CoA:cholesterol acyltransferase (ACAT). 7. Development of a series of substituted N-phenyl-N'-[(1-phenylcyclopentyl)methyl]ureas with enhanced hypocholesterolemic activity.
Journal of medicinal chemistry (1994-05-27)
B K Trivedi, T S Purchase, A Holmes, C E Augelli-Szafran, A D Essenburg, K L Hamelehle, R L Stanfield, R F Bousley, B R Krause
ABSTRACT
We recently described our initial structure-activity relationship (SAR) studies on a series of N-phenyl-N'-aralkyl- and N-phenyl-N'-(1-phenylcycloalkyl)ureas as inhibitors of acyl-CoA: cholesterol acyltransferase (ACAT). From this series of analogs, compound 1 (PD 129337) was identified as a potent inhibitor of ACAT with an IC50 value of 17 nM. It was also shown to dose-dependently lower plasma cholesterol in cholesterol-fed rats. However, further investigation led to the suggestion that this compound was poorly absorbed, due to a lack of efficacy when administered by gavage in an aqueous vehicle. To overcome this deficiency, we continued our SAR study on this novel series of ACAT inhibitors using an acute in vivo screen in which the compounds are administered to rats in an aqueous, CMC/Tween suspension vehicle. Modification of the N'-phenyl moiety by incorporating functional groups which were amenable to forming salts and/or polar groups to reduce lipophilicity led to the identification of several inhibitors which displayed excellent efficacy employing this protocol. Overall, substitution on the phenyl ring in the ortho, meta, or para positions led to inhibitors with only a slight decrease in potency in vitro compared to the parent unsubstituted compound. Bulkier groups in the para position tended to lower the ACAT inhibitory activity in vitro. Polar groups, such as carboxyl (33,34), lowered in vitro activity significantly, suggesting that polar-ionic interactions are disfavored for the enzyme activity. From this series, compound 28 was evaluated further in secondary in vivo screens. In a chronic cholesterol-fed rat model of hypercholesterolemia, compound 28 dose-dependently reduced nonHDL cholesterol and significantly elevated HDL cholesterol. It showed significantly greater aqueous solubility than the parent compound 1. However, it was shown to cause adrenal toxicity in guinea pigs. This led us to design a series of homologs (44-51) with increased basicity and lower lipophilicity. Some of these compounds were more potent ACAT inhibitors in vitro and demonstrated excellent hypocholesterolemic activity in vivo. Interestingly, compound 45, unlike 28, did not produce adrenal toxicity in guinea pigs and demonstrated excellent lipid-modulating activity in the chronic model of preestablished dyslipidemia in rats.
Sigma-Aldrich
ATR-101, ≥98% (HPLC)