MilliporeSigma
  • OSR1 regulates a subset of inward rectifier potassium channels via a binding motif variant.
OSR1 regulates a subset of inward rectifier potassium channels via a binding motif variant.
Proceedings of the National Academy of Sciences of the United States of America (2018-03-28)
Clinton A Taylor, Sung-Wan An, Sachith Gallolu Kankanamalage, Steve Stippec, Svetlana Earnest, Ashesh T Trivedi, Jonathan Zijiang Yang, Hamid Mirzaei, Chou-Long Huang, Melanie H Cobb
ABSTRACT
The with-no-lysine (K) (WNK) signaling pathway to STE20/SPS1-related proline- and alanine-rich kinase (SPAK) and oxidative stress-responsive 1 (OSR1) kinase is an important mediator of cell volume and ion transport. SPAK and OSR1 associate with upstream kinases WNK 1-4, substrates, and other proteins through their C-terminal domains which interact with linear R-F-x-V/I sequence motifs. In this study we find that SPAK and OSR1 also interact with similar affinity with a motif variant, R-x-F-x-V/I. Eight of 16 human inward rectifier K+ channels have an R-x-F-x-V motif. We demonstrate that two of these channels, Kir2.1 and Kir2.3, are activated by OSR1, while Kir4.1, which does not contain the motif, is not sensitive to changes in OSR1 or WNK activity. Mutation of the motif prevents activation of Kir2.3 by OSR1. Both siRNA knockdown of OSR1 and chemical inhibition of WNK activity disrupt NaCl-induced plasma membrane localization of Kir2.3. Our results suggest a mechanism by which WNK-OSR1 enhance Kir2.1 and Kir2.3 channel activity by increasing their plasma membrane localization. Regulation of members of the inward rectifier K+ channel family adds functional and mechanistic insight into the physiological impact of the WNK pathway.
Sigma-Aldrich
WNK463, ≥98% (HPLC)