MilliporeSigma
  • Stable expression of α1-antitrypsin Portland in MDA-MB-231 cells increased MT1-MMP and MMP-9 levels, but reduced tumour progression.
Stable expression of α1-antitrypsin Portland in MDA-MB-231 cells increased MT1-MMP and MMP-9 levels, but reduced tumour progression.
Journal of cell communication and signaling (2017-08-30)
J A Willson, C A Muir, C L Evered, M A Cepeda, S Damjanovski
ABSTRACT
The membrane bound matrix metalloproteinase MT1-MMP plays roles in modulating cell movement, independent of its abilities to remodel the extracellular matrix. Unlike many MMPs, MT1-MMP is activated in the Golgi prior to secretion by a pro-protein convertase, primarily furin. Regulation of the activation of pro-MT1-MMP has been methodically investigated, as altering the level of the active protein has broad implications in both activating other pro-MMPs, including pro-MMP-2, and many subsequent remodelling events. Our previous work in MCF-7 cells has demonstrated that modest, and not extremely high, levels of active MT1-MMP manifests into altered cell morphology and movement. At this low but optimal amount of MT1-MMP protein, changes to MT1-MMP levels are always mirrored by MMP-9 and pERK levels, and always opposite to MMP-2 levels. In this study, stable expression of the furin inhibitor α1-antitrypsin Portland (α1-PDX) in MDA-MB-231 cells increased overall MT1-MMP levels, but cells maintained a 21% proportion of pro-MT1-MMP. The increase in MT1-MMP was mirrored by increases in MMP-9 and pERK, but a decrease in MMP-2. These changes were associated with increased NF-κB transcription. In vitro analysis showed that α1-PDX decreased cell protrusions and migration, and this manifested as decreased tumourigenesis when examined in vivo using a chick CAM assay.
Sigma-Aldrich
Anti-MT1-MMP Antibody, hinge region, from rabbit, purified by affinity chromatography