MilliporeSigma
  • Effects of Zn(II) complex with vitamins C and U, and carnitine on metabolic syndrome model rats.
Effects of Zn(II) complex with vitamins C and U, and carnitine on metabolic syndrome model rats.
Metallomics : integrated biometal science (2011-06-28)
Kinuyo Matsumoto, Nao Motoyasu, Kyoko Sera, Takako Fujii, Yutaka Yoshikawa, Hiroyuki Yasui, Hiroshi Taniguchi, Naemi Kajiwara
ABSTRACT
The insulinomimetic activity of a Zn(ii) complex is reported. The effects of the Zn(ii) complex with ascorbic acid (Vitamin C; VC), methylmethionine sulfonium chloride (Vitamin U; VU) and l-carnitine were assessed in diet-induced metabolic syndrome model rats. Zn(VU)(2)Cl(2) and Zn(VC)Cl(2) were suggested to be useful supplementary materials for preventing metabolic syndrome by reducing visceral adipose tissues or accelerating blood fluidity.