MilliporeSigma
  • N6-methyladenosine ALKBH5 promotes non-small cell lung cancer progress by regulating TIMP3 stability.
N6-methyladenosine ALKBH5 promotes non-small cell lung cancer progress by regulating TIMP3 stability.
Gene (2020-01-14)
Zhiyuan Zhu, Qi Qian, Xia Zhao, Liang Ma, Ping Chen
ABSTRACT
Mounting evidence demonstrates that N6-methyladenosine (m6A) play critical roles of m6A in the epigenetic regulation, especially for human cancer. The m6A modification is installed by methyltransferase and erased demethylases, leading to the significant modification for gene expression and cell fate. Here, we investigated the biological roles and mechanism of demethylase alkylation repair homolog protein 5 (ALKBH5) in the non-small cell lung cancer (NSCLC). Results revealed that ALKBH5 was ectopically up-regulated in the NSCLC tissue and cells, and closely correlated with the poor prognosis. Functionally, ALKBH5 promoted the proliferation and reduced apoptosis of NSCLC cells in vitro, and knockdown of ALKBH5 repressed the tumor growth in vivo. Mechanistically, RNA immunoprecipitation sequencing (RIP-Seq) revealed that ALKBH5 targeted the TIMP3. Moreover, ALKBH5 repressed TIMP3 mRNA stability and protein production. In conclusion, the present research confirmed the ALKBH5/TIMP3 pathway in the NSCLC oncogenesis progress, providing a novel insight for the epitranscriptome and potential therapeutic target for NSCLC.
Sigma-Aldrich
MISSION® esiRNA, targeting human ALKBH5
Pricing and availability is not currently available.