MilliporeSigma
  • E3 ligase Herc4 regulates Hedgehog signalling through promoting Smoothened degradation.
E3 ligase Herc4 regulates Hedgehog signalling through promoting Smoothened degradation.
Journal of molecular cell biology (2019-03-30)
Weirong Jiang, Xia Yao, Zhaoliang Shan, Wenting Li, Yuxue Gao, Qing Zhang
ABSTRACT
Hedgehog (Hh) signalling plays conserved roles in controlling embryonic development; its dysregulation causes many diseases including cancers. The G protein-coupled receptor Smoothened (Smo) is the key signal transducer of the Hh pathway, whose posttranslational regulation has been shown to be critical for its accumulation and activation. Ubiquitination has been reported an essential posttranslational regulation of Smo. Here, we identify a novel E3 ligase of Smo, Herc4, which binds to Smo, and regulates Hh signalling by controlling Smo ubiquitination and degradation. Interestingly, our data suggest that Herc4-mediated Smo degradation is regulated by Hh in PKA-primed phosphorylation-dependent and independent manners.
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine, without calcium chloride and sodium bicarbonate, powder, suitable for insect cell culture
Pricing and availability is not currently available.