MilliporeSigma
  • Procedure for the Screening of Eggs and Egg Products to Detect Oxolonic Acid, Ciprofloxacin, Enrofloxacin, and Sarafloxacin Using Micellar Liquid Chromatography.
Procedure for the Screening of Eggs and Egg Products to Detect Oxolonic Acid, Ciprofloxacin, Enrofloxacin, and Sarafloxacin Using Micellar Liquid Chromatography.
Antibiotics (Basel, Switzerland) (2019-11-17)
Juan Peris-Vicente, Daniel García-Ferrer, Pooja Mishra, Jaume Albiol-Chiva, Abhilasha Durgbanshi, Samuel Carda-Broch, Devasish Bose, Josep Esteve-Romero
ABSTRACT
A method based on micellar liquid chromatography was developed to determine oxolinic acid, ciprofloxacin, enrofloxacin, and sarafloxacin in eggs and egg products. The antimicrobial drugs were obtained in a micellar solution which was directly injected. The analytes were resolved using a C18 column and a mobile phase of 0.05 M sodium dodecyl sulfate-7.5% 1-propanol-0.5% triethylamine, buffered at pH 3 with phosphate salt, running under the isocratic mode. The signal was monitored by fluorescence. Validation was successfully performed according to the EU Commission Decision 2002/657/EC in terms of specificity, calibration range (LOQ to 1 mg/kg), linearity (R2 > 0.9991), limit of detection and decision limit (0.01-0.05 mg/kg), limit of quantification (0.025-0.150 mg/kg), detection capability (<0.4 times decision limit), trueness (-14.2% to +9.8%), precision (<14.0%), robustness, and stability. The procedure was environmentally friendly, safe, easy-to-conduct, inexpensive, and had a high sample throughput, thus it is useful for routine analysis as a screening method in a laboratory for food residue control.
Sigma-Aldrich
2-Hydroxyquinoline, 98%