MilliporeSigma
  • Selective activation of Ca(2+)-dependent K+ channels by novel benzimidazolone.
Selective activation of Ca(2+)-dependent K+ channels by novel benzimidazolone.
European journal of pharmacology (1994-01-04)
S P Olesen, E Munch, P Moldt, J Drejer
ABSTRACT
Activators and blockers of specific ion channels are important pharmacological tools for characterizing ion channels and their influence on cell function. The large-conductance Ca(2+)-dependent K+ channel (BK channel) is blocked by peptides such as charybdotoxin and iberiotoxin, but no selective activator of the channel has been described. Here we report single-channel and whole-cell patch-clamp experiments on the specific activation of BK channels in aortic smooth muscle cells with a new heterocyclic molecule, NS 1619 (1-(2'-hydroxy-5'-trifluoromethylphenyl)-5-trifluoromethyl- 2(3H)benzimidazolone). The effect of NS 1619 on the BK channel was dose-dependent, resulting in a shift of the activation curve by up to -50 mV towards negative membrane potentials. The effect was fully reversible and was antagonized by charybdotoxin as well as by tetraethylammonium ions. The compound hyperpolarized the smooth muscle cells. NS 1619 is a selective and new type of K+ channel activator, which may significantly modulate cell excitability.
Sigma-Aldrich
NS 1619, powder