MilliporeSigma
  • Acetylation of alpha-fetoprotein promotes hepatocellular carcinoma progression.
Acetylation of alpha-fetoprotein promotes hepatocellular carcinoma progression.
Cancer letters (2019-12-08)
Junhui Xue, Zhengyi Cao, Yuning Cheng, Jiyin Wang, Yujuan Liu, Ruixiang Yang, Hui Li, Wei Jiang, Gang Li, Wenhui Zhao, Xiaowei Zhang
ABSTRACT
Alpha-fetoprotein (AFP) is a well-established biomarker for hepatocellular carcinoma (HCC). Here, we investigated the acetylation state of AFP in vivo. AFP acetylation was regulated by the acetyltransferase CBP and the deacetylase SIRT1. Acetylation of AFP at lysines 194, 211, and 242 increased the stability of AFP protein by decreasing its ubiquitination and proteasomal degradation. AFP acetylation promoted its oncogenic role by blocking binding to the phosphatase PTEN and the pro-apoptotic protein caspase-3, which increased signaling for proliferation, migration, and invasion and decreased apoptosis. High levels of acetylated AFP in HCC tissues were associated with HBV infection and correlated with poor prognosis and decreased patient survival. In HCC cells, hepatitis B virus X protein (HBx) and palmitic acid (PA) increased the level of acetylated AFP by disrupting SIRT1-mediated deacetylation. AFP acetylation plays an important role in HCC progression and provides a new potential prognostic marker and therapeutic target for HCC.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
p300, HAT Domain, Recombinant GST-fusion protein corresponding to amino acids 1066-1707 of human p300.