MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Targeted doxorubicin-containing luteinizing hormone-releasing hormone analogue AN-152 inhibits the growth of doxorubicin-resistant MX-1 human breast cancers.
Targeted doxorubicin-containing luteinizing hormone-releasing hormone analogue AN-152 inhibits the growth of doxorubicin-resistant MX-1 human breast cancers.
Clinical cancer research : an official journal of the American Association for Cancer Research (2003-09-25)
Ana M Bajo, Andrew V Schally, Gabor Halmos, Attila Nagy
ABSTRACT
The receptors for luteinizing hormone-releasing hormone receptor (LHRH-R) are found in >50% of human breast cancers. Doxorubicin (DOX) was linked to [D-Lys(6)]LHRH to form a cytotoxic conjugate, AN-152, which can be targeted to tumor cells expressing LHRH-R. We evaluated the effects of AN-152 on the estrogen-independent, DOX-resistant human mammary carcinoma line MX-1, xenografted into nude mice. Nude mice bearing MX-1 tumors were administered five i.v. injections of AN-152 or DOX at doses equivalent to 3 mg/kg DOX. Tumor growth was followed, and changes in the expression of LHRH-R on tumors were evaluated by radioreceptor assays, reverse transcription-PCR, and Western blotting. The effects of AN-152 on the expression of human epidermal growth factor receptor (HER)-2 were investigated. Because LHRH-R are coupled to various G proteins, which are involved in mitogenic signaling, we determined the outcome of treatment with AN-152 on the levels of mRNA for different G proteins. Treatment with AN-152 significantly (P < 0.05) decreased the final tumor volume to 978.56 +/- 176.85 mm(3), compared with the control tumors, which measured 2837.38 +/- 515.38 mm(3). Tumor doubling time was likewise significantly (P < 0.05) extended by AN-152 to 12.01 +/- 1.99 days from 6.45 +/- 0.36 days for the controls. Therapy with AN-152, but not with DOX, resulted in a significant decrease of LHRH-R levels on MX-1 tumors. The expression of mRNAs for HER-2, HER-3, Galpha(i2), and Galpha(11) and the levels of HER-2 and HER-3 proteins were also significantly reduced by AN-152. Cytotoxic LHRH analogue AN-152 could be considered for targeted chemotherapy of DOX-resistant breast cancers expressing LHRH-R.
Sigma-Aldrich
[D-Lys6]-LH-RH
Sigma-Aldrich
[D-Lys6]-LH-RH Free Acid
Sigma-Aldrich
[D-Phe2,D-Ala6]-LH-RH