MilliporeSigma
  • ARX788, a novel anti-HER2 antibody-drug conjugate, shows anti-tumor effects in preclinical models of trastuzumab emtansine-resistant HER2-positive breast cancer and gastric cancer.
ARX788, a novel anti-HER2 antibody-drug conjugate, shows anti-tumor effects in preclinical models of trastuzumab emtansine-resistant HER2-positive breast cancer and gastric cancer.
Cancer letters (2020-01-07)
Mark Barok, Vadim Le Joncour, Ana Martins, Jorma Isola, Marko Salmikangas, Pirjo Laakkonen, Heikki Joensuu
ABSTRACT
The majority of HER2-positive breast or gastric cancers treated with T-DM1 eventually show resistance to this agent. We compared the effects of T-DM1 and ARX788, a novel anti-HER2 antibody-drug conjugate, on cell growth and apoptosis in HER2-positive breast cancer and gastric cancer cell lines sensitive to T-DM1, gastric cancer cell lines resistant to T-DM1, HER2-negative breast cancer cell lines, and T-DM1-resistant xenograft models. ARX788 was effective in T-DM1-resistant in vitro and in vivo models of HER2-positive breast cancer and gastric cancer. ARX788 showed a pronounced growth inhibitory effect on all five HER2-positive cell lines tested, of which two gastric cancer cell lines had acquired resistance to T-DM1. ARX788 evoked more apoptotic events compared to T-DM1. While JIMT-1 and RN-87 xenograft tumors progressed on T-DM1 treatment, all such tumors responded to ARX788, and four out of the six JIMT-1 tumors and nine out of the twelve RN-87 tumors disappeared during the ARX788 treatment. Mice treated with ARX788 survived longer than those treated with T-DM1. The data support evaluation of ARX788 in patients with HER2-positive breast cancer or gastric cancer including cancers that progress during T-DM1 therapy.
Sigma-Aldrich
Anti-ABCC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1