MilliporeSigma
  • Purification and characterization of a novel prolyl aminopeptidase from Maitake (Grifola frondosa).
Purification and characterization of a novel prolyl aminopeptidase from Maitake (Grifola frondosa).
Bioscience, biotechnology, and biochemistry (2004-06-25)
Kazuyuki Hiwatashi, Kazuyuki Hori, Keitaro Takahashi, Akira Kagaya, Shunzo Inoue, Toshihiro Sugiyama, Saori Takahashi
ABSTRACT
We have found a novel prolyl aminopeptidase in Grifola frondosa. The enzyme was purified by DEAE-Sepharose CL-6B, Butyl-Toyopearl, Sephacryl S-100, and Mono-Q column chromatographies. The purified enzyme exists as a dimer and gives high activity toward L-proline-p-nitroanilide. The enzyme was strongly inhibited by p-chloromercuribenzoic acid and iodoacetic acid and markedly inhibited by phenylmethylsulfonyl fluoride and arphamenin A.
Sigma-Aldrich
L-Proline p-nitroanilide trifluoroacetate salt, prolyl aminopeptidase substrate
Pricing and availability is not currently available.