MilliporeSigma
  • Bevacizumab potentiates chemotherapeutic effect on T-leukemia/lymphoma cells by direct action on tumor endothelial cells.
Bevacizumab potentiates chemotherapeutic effect on T-leukemia/lymphoma cells by direct action on tumor endothelial cells.
Haematologica (2011-02-19)
Li Wang, Wen-Yu Shi, Fan Yang, Wei Tang, Guillaume Gapihan, Mariana Varna, Zhi-Xiang Shen, Sai-Juan Chen, Christophe Leboeuf, Anne Janin, Wei-Li Zhao
ABSTRACT
Vascular endothelial growth factor-A, an angiogenesis stimulator expressed on both tumor endothelial and malignant T cells, is involved in tumor progression in T-leukemia/lymphoma. Here, we assessed the impact of therapeutic vascular endothelial growth factor-A blockade on tumor-endothelial cell interaction and on tumor progression. In a murine xenograft T-leukemia/lymphoma model, combined bevacizumab (monoclonal antibody against vascular endothelial growth factor-A) with doxorubicin, compared with doxorubicin alone, significantly delayed tumor growth and induced prevalence of tumor cell apoptosis over mitosis. More importantly, the combined treatment induced endothelial cell swelling, microvessel occlusions, and tumor necrosis. In vitro, co-culture of endothelial cells with T-leukemia/lymphoma cells showed that doxorubicin induced expression of intracellular cell adhesion molecule-1, provided endothelial and malignant T cells were in direct contact. This was abrogated by bevacizumab treatment with doxorubicin. Taken together, bevacizumab enhances the chemotherapeutic effect on T-leukemia/lymphoma cells. Directly targeting tumor endothelial cells might be a promising therapeutic strategy to counteract tumor progression in T-cell malignancies.
Sigma-Aldrich
Anti-CD54 Antibody, extracellular, clone 84H10, clone 84H10, Chemicon®, from mouse