MilliporeSigma
Mechanism of thioflavin T binding to amyloid fibrils.
Journal of structural biology (2005-08-30)
Ritu Khurana, Chris Coleman, Cristian Ionescu-Zanetti, Sue A Carter, Vinay Krishna, Rajesh K Grover, Raja Roy, Shashi Singh
ABSTRACT
Thioflavin T is a benzothiazole dye that exhibits enhanced fluorescence upon binding to amyloid fibrils and is commonly used to diagnose amyloid fibrils, both ex vivo and in vitro. In aqueous solutions, thioflavin T was found to exist as micelles at concentrations commonly used to monitor fibrils by fluorescence assay ( approximately 10-20 microM). Specific conductivity changes were measured at varying concentration of thioflavin T and the critical micellar concentration was calculated to be 4.0+/-0.5 microM. Interestingly, changes in the fluorescence excitation and emission of thioflavin T were also dependent on the micelle formation. The thioflavin T micelles of 3 nm diameter were directly visualized using atomic force microscopy, and bound thioflavin T micelles were observed along the fibril length for representative fibrils. Increasing concentration of thioflavin T above the critical micellar concentration shows increased numbers of micelles bound along the length of the amyloid fibrils. Thioflavin T micelles were disrupted at low pH as observed by atomic force microscopy and fluorescence enhancement upon binding of thioflavin T to amyloid fibrils also reduced by several-fold upon decreasing the pH to below 3. This suggests that positive charge on the thioflavin T molecule has a role in its micelle formation that then bind the amyloid fibrils. Our data suggests that the micelles of thioflavin T bind amyloid fibrils leading to enhancement of fluorescence emission.
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Pricing and availability is not currently available.