MilliporeSigma
  • Beyond the brain-Peripheral kisspeptin signaling is essential for promoting endometrial gland development and function.
Beyond the brain-Peripheral kisspeptin signaling is essential for promoting endometrial gland development and function.
Scientific reports (2016-07-02)
Silvia León, Daniela Fernandois, Alexandra Sull, Judith Sull, Michele Calder, Kanako Hayashi, Moshmi Bhattacharya, Stephen Power, George A Vilos, Angelos G Vilos, Manuel Tena-Sempere, Andy V Babwah
ABSTRACT
Uterine growth and endometrial gland formation (adenogenesis) and function, are essential for fertility and are controlled by estrogens and other regulators, whose nature and physiological relevance are yet to be elucidated. Kisspeptin, which signals via Kiss1r, is essential for fertility, primarily through its central control of the hypothalamic-pituitary-ovarian axis, but also likely through peripheral actions. Using genetically modified mice, we addressed the contributions of central and peripheral kisspeptin signaling in regulating uterine growth and adenogenesis. Global ablation of Kiss1 or Kiss1r dramatically suppressed uterine growth and almost fully prevented adenogenesis. However, while uterine growth was fully rescued by E2 treatment of Kiss1(-/-) mice and by genetic restoration of kisspeptin signaling in GnRH neurons in Kiss1r(-/-) mice, functional adenogenesis was only marginally restored. Thus, while uterine growth is largely dependent on ovarian E2-output via central kisspeptin signaling, peripheral kisspeptin signaling is indispensable for endometrial adenogenesis and function, essential aspects of reproductive competence.
Sigma-Aldrich
Anti-Osteopontin Antibody, serum, from rabbit
Pricing and availability is not currently available.