MilliporeSigma
  • O-GlcNAcylation of the Signaling Scaffold Protein, GNB2L1 Promotes its Degradation and Increases Metastasis of Gastric Tumours.
O-GlcNAcylation of the Signaling Scaffold Protein, GNB2L1 Promotes its Degradation and Increases Metastasis of Gastric Tumours.
Biochemical and biophysical research communications (2016-08-18)
Shimeng Cheng, Jie Ren, Lina Su, Jianlan Liu, Qingmei Liu, Jing Zhou, Xiaolu Ye, Ningwen Zhu
ABSTRACT
GNB2L1 is an intercellular scaffold protein of the Trp-Asp (WD) repeat protein family, and has been reported to play suppressive roles in the progression of gastric cancer. However, the regulatory mechanisms of GNB2L1 in gastric cancer still remain largely elusive. In the present study, we found that OGT was capable to interact with GNB2L1 directly and modify GNB2L1 with O-GlcNAcylation in gastric cancer, and this O-GlcNAcylation hindered the inhibition of GNB2L1 on migration of gastric cancer cells. Moreover, O-GlcNAcylation regulated the degradation instead of the synthesis of GNB2L1 in gastric cancer, and our data suggested the O-GlcNAcylation on GNB2L1 influenced its stability directly. In addition, the clinical data revealed the negative correlation of the protein level instead of the mRNA level of GNB2L1 with OGT expression, and showed that OGT reversed the inhibition of GNB2L1 on metastasis, and worsened the prognosis of GNB2L1(High) patients. In summary, this study indicated the O-GlcNAcylation on GNB2L1 reversed its inhibition on gastric tumour metastasis via promoting its degradation.
Sigma-Aldrich
ST045849, ≥98% (HPLC)
Pricing and availability is not currently available.