MilliporeSigma
  • Characterization of the Streptococcus mutans P1 epitope recognized by immunomodulatory monoclonal antibody 6-11A.
Characterization of the Streptococcus mutans P1 epitope recognized by immunomodulatory monoclonal antibody 6-11A.
Infection and immunity (2004-07-24)
Nikki R Rhodin, Jenny M Cutalo, Kenneth B Tomer, William P McArthur, L Jeannine Brady
ABSTRACT
Monoclonal antibody (MAb) 6-11A directed against Streptococcus mutans surface adhesin P1 was shown previously to influence the mucosal immunogenicity of this organism in BALB/c mice. The specificity of anti-P1 serum immunoglobulin G (IgG) and secretory IgA antibodies and the subclass distribution of anti-P1 serum IgG antibodies were altered, and the ability of elicited serum antibodies to inhibit S. mutans adherence in vitro was in certain cases increased. MAb 6-11A is known to recognize an epitope dependent on the presence of the proline-rich region of the protein, although it does not bind directly to the isolated P-region domain. In this report, we show that MAb 6-11A recognizes a complex discontinuous epitope that requires the simultaneous presence of the alanine-rich repeat domain (A-region) and the P-region. Formation of the core epitope requires the interaction of these segments of P1. Residues amino terminal to the A-region also contributed to recognition by MAb 6-11A but were not essential for binding. Characterization of the MAb 6-11A epitope will enable insight into potential mechanisms of immunomodulation and broaden our understanding of the tertiary structure of P1.