MilliporeSigma
  • Myxoma virus M013 protein antagonizes NF-κB and inflammasome pathways via distinct structural motifs.
Myxoma virus M013 protein antagonizes NF-κB and inflammasome pathways via distinct structural motifs.
The Journal of biological chemistry (2019-04-04)
Rekha R Garg, Cody B Jackson, Masmudur M Rahman, Amir R Khan, Alfred S Lewin, Grant McFadden
ABSTRACT
Among the repertoire of immunoregulatory proteins encoded by myxoma virus, M013 is a viral homologue of the viral pyrin domain-only protein (vPOP) family. In myeloid cells, M013 protein has been shown to inhibit both the inflammasome and NF-κB signaling pathways by direct binding to ASC1 and NF-κB1, respectively. In this study, a three-dimensional homology model of the M013 pyrin domain (PYD) was built based on similarities to known PYD structures. A distinctive feature of the deduced surface electrostatic map of the M013 PYD is the presence of a negatively region consisting of numerous aspartate and glutamate residues in close proximity. Single-site mutations of aspartate and glutamate residues reveal their role in interactions with ASC-1. The biological significance of charge complementarity in the M013-ASC-1 interaction was further confirmed by functional assays of caspase-1 activation and subsequent secretion of cytokines. M013 also has a unique 33-residue C-terminal tail that follows the N-terminal PYD, and it is enriched in positively charged residues. Deletion of the tail of M013 significantly inhibited the interactions between M013 and NF-κB1, thus compromising the ability of the viral protein to suppress the secretion of pro-inflammatory cytokines. These results demonstrate that vPOP M013 exploits distinct structural motifs to regulate both the inflammasome and NF-κB pathways.
Sigma-Aldrich
Anti-ASC Antibody, Chemicon®, from rabbit