MilliporeSigma
  • DJ-1 can form β-sheet structured aggregates that co-localize with pathological amyloid deposits.
DJ-1 can form β-sheet structured aggregates that co-localize with pathological amyloid deposits.
Neurobiology of disease (2019-11-02)
Katalin Solti, Wei-Li Kuan, Balázs Fórizs, Gergely Kustos, Judith Mihály, Zoltán Varga, Balázs Herberth, Éva Moravcsik, Róbert Kiss, Manuela Kárpáti, Anna Mikes, Yanyan Zhao, Tímea Imre, Jean-Christophe Rochet, Franklin Aigbirhio, Caroline H Williams-Gray, Roger A Barker, Gergely Tóth
ABSTRACT
The loss of native function of the DJ-1 protein has been linked to the development of Parkinson's (PD) and other neurodegenerative diseases. Here we show that DJ-1 aggregates into β-sheet structured soluble and fibrillar aggregates in vitro under physiological conditions and that this process is promoted by the oxidation of its catalytic Cys106 residue. This aggregation resulted in the loss of its native biochemical glyoxalase function and in addition oxidized DJ-1 aggregates were observed to localize within Lewy bodies, neurofibrillary tangles and amyloid plaques in human PD and Alzheimer's (AD) patients' post-mortem brain tissue. These findings suggest that the aggregation of DJ-1 may be a critical player in the development of the pathology of PD and AD and demonstrate that loss of DJ-1 function can happen through DJ-1 aggregation. This could then contribute to AD and PD disease onset and progression.
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-oxDJ-1 Antibody (Cys106), clone M149, clone M149, from mouse
Pricing and availability is not currently available.