MilliporeSigma
  • Spell Checking Nature: Versatility of CRISPR/Cas9 for Developing Treatments for Inherited Disorders.
Spell Checking Nature: Versatility of CRISPR/Cas9 for Developing Treatments for Inherited Disorders.
American journal of human genetics (2015-12-22)
Daria Wojtal, Dwi U Kemaladewi, Zeenat Malam, Sarah Abdullah, Tatianna W Y Wong, Elzbieta Hyatt, Zahra Baghestani, Sergio Pereira, James Stavropoulos, Vincent Mouly, Kamel Mamchaoui, Francesco Muntoni, Thomas Voit, Hernan D Gonorazky, James J Dowling, Michael D Wilson, Roberto Mendoza-Londono, Evgueni A Ivakine, Ronald D Cohn
ABSTRACT
Clustered regularly interspaced short palindromic repeat (CRISPR) has arisen as a frontrunner for efficient genome engineering. However, the potentially broad therapeutic implications are largely unexplored. Here, to investigate the therapeutic potential of CRISPR/Cas9 in a diverse set of genetic disorders, we establish a pipeline that uses readily obtainable cells from affected individuals. We show that an adapted version of CRISPR/Cas9 increases the amount of utrophin, a known disease modifier in Duchenne muscular dystrophy (DMD). Furthermore, we demonstrate preferential elimination of the dominant-negative FGFR3 c.1138G>A allele in fibroblasts of an individual affected by achondroplasia. Using a previously undescribed approach involving single guide RNA, we successfully removed large genome rearrangement in primary cells of an individual with an X chromosome duplication including MECP2. Moreover, removal of a duplication of DMD exons 18-30 in myotubes of an individual affected by DMD produced full-length dystrophin. Our findings establish the far-reaching therapeutic utility of CRISPR/Cas9, which can be tailored to target numerous inherited disorders.
Sigma-Aldrich
Anti-Dystrophin Antibody, mid-rod, clone 6D3, culture supernatant, clone 6D3, Chemicon®