MilliporeSigma
  • Schwann cells secrete extracellular vesicles to promote and maintain the proliferation and multipotency of hDPCs.
Schwann cells secrete extracellular vesicles to promote and maintain the proliferation and multipotency of hDPCs.
Cell proliferation (2017-07-18)
Ziyue Li, Yan Liang, Kuangwu Pan, Hui Li, Mei Yu, Weihua Guo, Guoqing Chen, Weidong Tian
ABSTRACT
Schwann cells (SCs) are the principal glial cells in peripheral nerve system, involved in neuropathies with great regenerative potential. Dental pulp cells have been reported to maintain neurogenic potential. In contrast, the regulatory role of SCs on human dental pulp cells (hDPCs) development remains undefined. SC secretion and SC-derived extracellular vesicles (EVs) were collected and used to treat hDPCs; and proliferation and multiple differentiation of hDPCs were detected after EVs treatments. Finally, we analysed the proteomes of SC-EVs and SCs through mass spectrum. In this study, we found SC secretion showed a predominantly regulatory role on the development of hDPCs. Further, we identified EVs from SC secretion with similar function as SC secretion in regulating hDPCs proliferation and multipotency. And expression of transcription factor Oct4 was upregulated after treatment of both SC secretion and EVs, as well as Sox2 and Nanog. We detected abundant enrichment of Oct4 in EVs, which might be responsible for the upregulation of stem cell-related genes in hDPCs. Through proteome and western blot analysis, we found enriched TGFβs in EVs, indicating that accelerated hDPCs proliferation may be mediated by activated TGFβ-Samd and TGFβ-MAPK signalling. In summary, our study sheds light on critical regulatory ability of SC-derived EVs on hDPCs proliferation and multipotency, suggesting great implications for seeding cells used in tissue engineering.
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone RCK107, clone RCK107, Chemicon®, from mouse