MilliporeSigma
  • Use of 2,6-dihydroxyacetophenone for analysis of fragile peptides, disulphide bonding and small proteins by matrix-assisted laser desorption/ionization.
Use of 2,6-dihydroxyacetophenone for analysis of fragile peptides, disulphide bonding and small proteins by matrix-assisted laser desorption/ionization.
Rapid communications in mass spectrometry : RCM (1996-01-01)
J J Gorman, B L Ferguson, T B Nguyen
ABSTRACT
Several peptides were shown to undergo fragmentation during matrix-assisted laser desorption/ionization time-of-flight mass spectrometry to a degree which complicated their analysis using alpha-cyano-4-hydroxycinnamic acid (CHCA) as a matrix, even at threshold laser irradiance. These peptides included synthetic peptides, peptides isolated from viral proteins and a phosphopeptide from beta-casein (residues 33-48). The excessive fragmentation occurred usually as a post-source phenomenon; however, in-source fragmentation was also observed. The combined effects of in-source and post-source fragmentation of one peptide studied led to a failure to observe the protonated molecule of this peptide in reflector mode analysis. The phosphopeptide studied exhibited a high degree of beta-elimination of phosphate. It was demonstrated that the fragility exhibited by these peptides in CHCA, including beta-elimination of phosphate from serine, was not evident with a matrix comprising 2,6-dihydroxyacetophenone (DHAP) and di-ammonium hydrogen citrate (DAHC). The DHAP/DAHC matrix was also adapted for direct analysis of peptides from an acidic reducing milieu containing tris(2-carboxyethyl)phosphine. The molecular weight of equine cytochrome c was determined with a relatively high degree of accuracy (experimental M(r) = 12360.2 +/- 1.4 Da compared to the theoretical M(r) = 12360.09 Da) using DHAP/DAHC as a matrix for reflector mode analysis.
Product Description
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
Sigma-Aldrich
2′,6′-Dihydroxyacetophenone, 97%