MilliporeSigma
  • Signaling through the interleukin 2 receptor beta chain activates a STAT-5-like DNA-binding activity.
Signaling through the interleukin 2 receptor beta chain activates a STAT-5-like DNA-binding activity.
Proceedings of the National Academy of Sciences of the United States of America (1995-08-01)
S L Gaffen, S Y Lai, W Xu, F Gouilleux, B Groner, M A Goldsmith, W C Greene
ABSTRACT
To explore the possible involvement of STAT factors ("signal transducers and activators of transcription") in the interleukin 2 receptor (IL-2R) signaling cascade, murine HT-2 cells expressing chimeric receptors composed of the extracellular domain of the erythropoietin receptor fused to the cytoplasmic domains of the IL-2R beta or -gamma c chains were prepared. Erythropoietin or IL-2 activation of these cells resulted in rapid nuclear expression of a DNA-binding activity that reacted with select STAT response elements. Based on reactivity with specific anti-STAT antibodies, this DNA-binding activity was identified as a murine homologue of STAT-5. Induction of nuclear expression of this STAT-5-like factor was blocked by the addition of herbimycin A, a tyrosine kinase inhibitor, but not by rapamycin, an immunophilin-binding antagonist of IL-2-induced proliferation. The IL-2R beta chain appeared critical for IL-2-induced activation of STAT-5, since a mutant beta chain lacking all cytoplasmic tyrosine residues was incapable of inducing this DNA binding. In contrast, a gamma c mutant lacking all of its cytoplasmic tyrosine residues proved fully competent for the induction of STAT-5. Physical binding of STAT-5 to functionally important tyrosine residues within IL-2R beta was supported by the finding that phosphorylated, but not nonphosphorylated, peptides corresponding to sequences spanning Y392 and Y510 of the IL-2R beta tail specifically inhibited STAT-5 DNA binding.
Sigma-Aldrich
Monoclonal Anti-Phosphothreonine antibody produced in mouse, clone PTR-8, ascites fluid