MilliporeSigma
  • Kupffer Cell-Derived Tnf Triggers Cholangiocellular Tumorigenesis through JNK due to Chronic Mitochondrial Dysfunction and ROS.
Kupffer Cell-Derived Tnf Triggers Cholangiocellular Tumorigenesis through JNK due to Chronic Mitochondrial Dysfunction and ROS.
Cancer cell (2017-06-14)
Detian Yuan, Shan Huang, Emanuel Berger, Lei Liu, Nina Gross, Florian Heinzmann, Marc Ringelhan, Tracy O Connor, Mira Stadler, Michael Meister, Julia Weber, Rupert Öllinger, Nicole Simonavicius, Florian Reisinger, Daniel Hartmann, Rüdiger Meyer, Maria Reich, Marco Seehawer, Valentina Leone, Bastian Höchst, Dirk Wohlleber, Simone Jörs, Marco Prinz, Duncan Spalding, Ulrike Protzer, Tom Luedde, Luigi Terracciano, Matthias Matter, Thomas Longerich, Percy Knolle, Thomas Ried, Verena Keitel, Fabian Geisler, Kristian Unger, Einat Cinnamon, Eli Pikarsky, Norbert Hüser, Roger J Davis, Darjus F Tschaharganeh, Roland Rad, Achim Weber, Lars Zender, Dirk Haller, Mathias Heikenwalder
ABSTRACT
Intrahepatic cholangiocarcinoma (ICC) is a highly malignant, heterogeneous cancer with poor treatment options. We found that mitochondrial dysfunction and oxidative stress trigger a niche favoring cholangiocellular overgrowth and tumorigenesis. Liver damage, reactive oxygen species (ROS) and paracrine tumor necrosis factor (Tnf) from Kupffer cells caused JNK-mediated cholangiocellular proliferation and oncogenic transformation. Anti-oxidant treatment, Kupffer cell depletion, Tnfr1 deletion, or JNK inhibition reduced cholangiocellular pre-neoplastic lesions. Liver-specific JNK1/2 deletion led to tumor reduction and enhanced survival in Akt/Notch- or p53/Kras-induced ICC models. In human ICC, high Tnf expression near ICC lesions, cholangiocellular JNK-phosphorylation, and ROS accumulation in surrounding hepatocytes are present. Thus, Kupffer cell-derived Tnf favors cholangiocellular proliferation/differentiation and carcinogenesis. Targeting the ROS/Tnf/JNK axis may provide opportunities for ICC therapy.
Sigma-Aldrich
Anti-E-cadherin Antibody, clone DECMA-1, clone Decma-1, from rat