MilliporeSigma
  • Requirement of the p130CAS-Crk coupling for metastasis suppressor KAI1/CD82-mediated inhibition of cell migration.
Requirement of the p130CAS-Crk coupling for metastasis suppressor KAI1/CD82-mediated inhibition of cell migration.
The Journal of biological chemistry (2003-05-10)
Xin A Zhang, Bo He, Bin Zhou, Li Liu
ABSTRACT
KAI1/CD82 protein is a member of the tetraspanin superfamily and has been rediscovered as a cancer metastasis suppressor. The mechanism of KAI1/CD82-mediated suppression of cancer metastasis remains to be established. In this study, we found that migration of the metastatic prostate cancer cell line Du145 was substantially inhibited when KAI1/CD82 was expressed. The expression of focal adhesion kinase (FAK) and Lyn, a Src family tyrosine kinase and substrate of FAK, was up-regulated at both RNA and protein levels upon KAI1/CD82 expression. The activation of FAK and Lyn, however, remained unchanged in Du145-KAI1/CD82 cells. As a downstream target of FAK-Lyn signaling, the p130CAS (Crk-associated substrate) protein was decreased upon the expression of KAI1/CD82. Consequently, less p130CAS-CrkII complex, which functions as a "molecular switch" in cell motility, was formed in Du145-KAI1/CD82 cells. To confirm that the p130CAS-CrkII complex is indeed important for the motility inhibition by KAI1/CD82, overexpression of p130CAS in Du145-KAI1/CD82 cells increased the formation of p130CAS-CrkII complex and largely reversed the KAI1/CD82-mediated inhibition of cell motility. Taken together, our studies indicate the following: 1) signaling of FAK-Lyn-p130CAS-CrkII pathway is altered in KAI1/CD82-expressing cells, and 2) p130CAS-CrkII coupling is required for KAI1/CD82-mediated suppression of cell motility.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse