MilliporeSigma
  • Long-term Tumor Adaptation after Radiotherapy: Therapeutic Implications for Targeting Integrins in Prostate Cancer.
Long-term Tumor Adaptation after Radiotherapy: Therapeutic Implications for Targeting Integrins in Prostate Cancer.
Molecular cancer research : MCR (2018-07-26)
Iris Eke, Adeola Y Makinde, Molykutty J Aryankalayil, Jessica L Reedy, Deborah E Citrin, Sunita Chopra, Mansoor M Ahmed, C Norman Coleman
ABSTRACT
Adaptation of tumor cells to radiotherapy induces changes that are actionable by molecular targeted agents and immunotherapy. This report demonstrates that radiation-induced changes in integrin expression can be targeted 2 months later. Integrins are transmembrane cell adhesion molecules that are essential for cancer cell survival and proliferation. To analyze the short- and long-term effects of radiation on the integrin expression, prostate cancer cells (DU145, PC3, and LNCaP) were cultured in a 3D extracellular matrix and irradiated with either a single dose of radiation (2-10 Gy) or a multifractionated regimen (2-10 fractions of 1 Gy). Whole human genome microarrays, immunoblotting, immunoprecipitation assays, and immunofluorescence staining of integrins were performed. The results were confirmed in a prostate cancer xenograft model system. Interestingly, β1 and β4 integrins (ITGB1 and ITGB4) were upregulated after radiation in vitro and in vivo. This overexpression lasted for more than 2 months and was dose dependent. Moreover, radiation-induced upregulation of β1 and β4 integrin resulted in significantly increased tumor cell death after treatment with inhibitory antibodies. Combined, these findings indicate that long-term tumor adaptation to radiation can result in an increased susceptibility of surviving cancer cells to molecular targeted therapy due to a radiation-induced overexpression of the target. IMPLICATIONS: Radiation induces dose- and schedule-dependent adaptive changes that are targetable for an extended time; thus suggesting radiotherapy as a unique strategy to orchestrate molecular processes, thereby providing new radiation-drug treatment options within precision cancer medicine.
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-8, clone ASC-8, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin Beta1, clone AIIB2 (Azide Free) Antibody, clone AIIB2, 1 mg/mL, from rat
Pricing and availability is not currently available.