MilliporeSigma
  • Glia contribute to the purinergic modulation of inspiratory rhythm-generating networks.
Glia contribute to the purinergic modulation of inspiratory rhythm-generating networks.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2010-03-20)
Adrianne G Huxtable, Jennifer D Zwicker, Tucaauê S Alvares, Araya Ruangkittisakul, Xin Fang, Leanne B Hahn, Elena Posse de Chaves, Glen B Baker, Klaus Ballanyi, Gregory D Funk
ABSTRACT
Glia modulate neuronal activity by releasing transmitters in a process called gliotransmission. The role of this process in controlling the activity of neuronal networks underlying motor behavior is unknown. ATP features prominently in gliotransmission; it also contributes to the homeostatic ventilatory response evoked by low oxygen through mechanisms that likely include excitation of preBötzinger complex (preBötC) neural networks, brainstem centers critical for breathing. We therefore inhibited glial function in rhythmically active inspiratory networks in vitro to determine whether glia contribute to preBötC ATP sensitivity. Glial toxins markedly reduced preBötC responses to ATP, but not other modulators. Furthermore, since preBötC glia responded to ATP with increased intracellular Ca(2+) and glutamate release, we conclude that glia contribute to the ATP sensitivity of preBötC networks, and possibly the hypoxic ventilatory response. Data reveal a role for glia in signal processing within brainstem motor networks that may be relevant to similar networks throughout the neuraxis.
Sigma-Aldrich
Anti-Neurofilament 160 kDa Antibody, clone NN18, clone NN18, Chemicon®, from mouse